Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACTTACCCCATTGAGGCTGCCCAA[A/G]TCCTTCACTAAATGCTCATCCGTAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613023 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEP170 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CEP170 - centrosomal protein 170 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042404.1 | 585 | Silent Mutation | GAC,GAT | D,D 59 | NP_001035863.1 | |
NM_001042405.1 | 585 | Silent Mutation | GAC,GAT | D,D 59 | NP_001035864.1 | |
NM_014812.2 | 585 | Silent Mutation | GAC,GAT | D,D 59 | NP_055627.2 | |
XM_006711843.3 | 585 | Silent Mutation | GAC,GAT | D,D 59 | XP_006711906.1 | |
XM_011544334.2 | 585 | Silent Mutation | GAC,GAT | D,D 59 | XP_011542636.1 | |
XM_011544335.2 | 585 | Silent Mutation | GAC,GAT | D,D 59 | XP_011542637.1 | |
XM_011544336.2 | 585 | Silent Mutation | GAC,GAT | D,D 59 | XP_011542638.1 | |
XM_011544337.2 | 585 | Silent Mutation | GAC,GAT | D,D 59 | XP_011542639.1 | |
XM_011544338.2 | 585 | Silent Mutation | GAC,GAT | D,D 59 | XP_011542640.1 | |
XM_011544339.2 | 585 | Silent Mutation | GAC,GAT | D,D 59 | XP_011542641.1 | |
XM_011544340.2 | 585 | Silent Mutation | GAC,GAT | D,D 59 | XP_011542642.1 | |
XM_011544341.2 | 585 | Silent Mutation | GAC,GAT | D,D 59 | XP_011542643.1 | |
XM_011544342.2 | 585 | Silent Mutation | GAC,GAT | D,D 59 | XP_011542644.1 | |
XM_011544343.2 | 585 | Silent Mutation | GAC,GAT | D,D 59 | XP_011542645.1 | |
XM_011544344.2 | 585 | Silent Mutation | GAC,GAT | D,D 59 | XP_011542646.1 | |
XM_017002932.1 | 585 | Silent Mutation | GAC,GAT | D,D 59 | XP_016858421.1 | |
XM_017002933.1 | 585 | Silent Mutation | GAC,GAT | D,D 59 | XP_016858422.1 | |
XM_017002934.1 | 585 | Silent Mutation | GAC,GAT | D,D 59 | XP_016858423.1 | |
XM_017002935.1 | 585 | Silent Mutation | GAC,GAT | D,D 59 | XP_016858424.1 | |
XM_017002936.1 | 585 | Silent Mutation | GAC,GAT | D,D 59 | XP_016858425.1 | |
XM_017002937.1 | 585 | Silent Mutation | GAC,GAT | D,D 59 | XP_016858426.1 | |
XM_017002938.1 | 585 | Silent Mutation | GAC,GAT | D,D 59 | XP_016858427.1 | |
XM_017002939.1 | 585 | Silent Mutation | GAC,GAT | D,D 59 | XP_016858428.1 | |
XM_017002940.1 | 585 | Silent Mutation | GAC,GAT | D,D 59 | XP_016858429.1 | |
XM_017002941.1 | 585 | Silent Mutation | GAC,GAT | D,D 59 | XP_016858430.1 | |
XM_017002942.1 | 585 | Silent Mutation | GAC,GAT | D,D 59 | XP_016858431.1 | |
XM_017002943.1 | 585 | Silent Mutation | GAC,GAT | D,D 59 | XP_016858432.1 | |
XM_017002944.1 | 585 | Silent Mutation | GAC,GAT | D,D 59 | XP_016858433.1 | |
XM_017002945.1 | 585 | Silent Mutation | GAC,GAT | D,D 59 | XP_016858434.1 | |
XM_017002946.1 | 585 | Silent Mutation | GAC,GAT | D,D 59 | XP_016858435.1 | |
XM_017002947.1 | 585 | Silent Mutation | GAC,GAT | D,D 59 | XP_016858436.1 | |
XM_017002948.1 | 585 | Silent Mutation | GAC,GAT | D,D 59 | XP_016858437.1 | |
XM_017002949.1 | 585 | Silent Mutation | GAC,GAT | D,D 59 | XP_016858438.1 | |
XM_017002950.1 | 585 | Silent Mutation | GAC,GAT | D,D 59 | XP_016858439.1 | |
XM_017002951.1 | 585 | Silent Mutation | GAC,GAT | D,D 59 | XP_016858440.1 |