Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCTCTCTCCTTCCATGTCACCTG[A/G]ATCGCATCCTTTTACAACCATTCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 188370 | ||||||||||||||||||||
Literature Links: |
CD1A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CD1A - CD1a molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320652.1 | 118 | Nonsense Mutation | TGA,TGG | *,W 20 | NP_001307581.1 | |
NM_001763.2 | 118 | Nonsense Mutation | TGA,TGG | *,W 31 | NP_001754.2 |