Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTGCCTGGAATTGAAGCCAGAGCC[A/G]ACCAAAGTAAGAAGCCCTTTTGAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
TEX35 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TEX35 - testis expressed 35 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001170722.1 | 196 | Silent Mutation | CCA,CCG | P,P 36 | NP_001164193.1 | |
NM_001170723.1 | 196 | Silent Mutation | CCA,CCG | P,P 28 | NP_001164194.1 | |
NM_001170724.1 | 196 | Silent Mutation | CCA,CCG | P,P 28 | NP_001164195.1 | |
NM_032126.4 | 196 | Silent Mutation | CCA,CCG | P,P 28 | NP_115502.2 |