Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACGGCCGTGCTAACAGTGGACTCG[A/G]AGGAGCCCAGCTTCAAGGCGGGGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
12 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 604835 | ||||||||||||||||||||
Literature Links: |
DUSP12 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DUSP12 - dual specificity phosphatase 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_007240.2 | 218 | Missense Mutation | AAG,GAG | K,E 62 | NP_009171.1 | |
XM_005244862.2 | 218 | UTR 5 | XP_005244919.1 |