Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGATGATAAATTGGAGTGAGAAATC[A/G]GATCTGAGGAGGTTCAAATGGGTAC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 610538 | ||||||||||||||||||||
Literature Links: |
UBE2T PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
UBE2T - ubiquitin conjugating enzyme E2 T | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001310326.1 | 341 | Nonsense Mutation | CGA,TGA | R,* 39 | NP_001297255.1 | |
NM_014176.3 | 341 | Nonsense Mutation | CGA,TGA | R,* 69 | NP_054895.1 |