Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGCGGCCTTCTCGCCCGGGCAGGA[A/G]GAGCCGCCGCGGCGCCGCGGCCGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
8 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606644 MIM: 600932 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IGSF8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IGSF8 - immunoglobulin superfamily member 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KCNJ9 - potassium voltage-gated channel subfamily J member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004983.2 | 281 | Silent Mutation | GAA,GAG | E,E 13 | NP_004974.2 | |
XM_017001247.1 | 281 | Silent Mutation | GAA,GAG | E,E 70 | XP_016856736.1 |