Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCCACAGAGGCATCATGGAGGGTC[C/G]CCGGGGATGGCTGGTGCTCTGTGTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 120550 MIM: 120575 | ||||||||||||||||||||
Literature Links: |
C1QA PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C1QA - complement C1q A chain | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015991.2 | 96 | Missense Mutation | CCC,CGC | P,R 4 | NP_057075.1 |
C1QC - complement C1q C chain | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6127 - microRNA 6127 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |