Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGGATGGCGGAAGTGGAGGCGCCGA[A/C]GGCGGCCGAGACGGACATGAAGCAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
10 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
ACTG1P20 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ACTG1P20 - actin gamma 1 pseudogene 20 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMEM222 - transmembrane protein 222 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032125.2 | 136 | Missense Mutation | AAG,ACG | K,T 28 | NP_115501.2 |