Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGATGAACACCGCCAAGACCGTGC[A/C]CACCATCCACAGCGCCCGGCGCTTG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 173393 | ||||||||||||||||||||
Literature Links: |
PTAFR PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PTAFR - platelet activating factor receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000952.4 | 1038 | Missense Mutation | GGC,TGC | G,C 235 | NP_000943.1 | |
NM_001164721.1 | 1038 | Missense Mutation | GGC,TGC | G,C 235 | NP_001158193.1 | |
NM_001164722.2 | 1038 | Missense Mutation | GGC,TGC | G,C 235 | NP_001158194.1 | |
NM_001164723.2 | 1038 | Missense Mutation | GGC,TGC | G,C 235 | NP_001158195.1 |