Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATTTTTCTTTCTAAGCTCAGTTT[C/G]CAGATCTAATATTTTAATTTGTAAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
MIR7856 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MIR7856 - microRNA 7856 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ODF2L - outer dense fiber of sperm tails 2 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007022.2 | 2155 | Missense Mutation | CAA,GAA | Q,E 612 | NP_001007023.2 | |
NM_001184765.1 | 2155 | Missense Mutation | CAA,GAA | Q,E 559 | NP_001171694.1 | |
NM_001184766.1 | 2155 | Missense Mutation | CAA,GAA | Q,E 530 | NP_001171695.1 | |
NM_020729.2 | 2155 | Missense Mutation | CAA,GAA | Q,E 596 | NP_065780.2 | |
XM_005271054.3 | 2155 | Missense Mutation | CAA,GAA | Q,E 583 | XP_005271111.1 | |
XM_011541815.2 | 2155 | Missense Mutation | CAA,GAA | Q,E 612 | XP_011540117.1 | |
XM_011541819.2 | 2155 | Missense Mutation | CAA,GAA | Q,E 612 | XP_011540121.1 | |
XM_011541821.2 | 2155 | Missense Mutation | CAA,GAA | Q,E 559 | XP_011540123.1 | |
XM_011541823.2 | 2155 | Intron | XP_011540125.1 | |||
XM_011541824.2 | 2155 | Intron | XP_011540126.1 | |||
XM_017001876.1 | 2155 | Missense Mutation | CAA,GAA | Q,E 612 | XP_016857365.1 | |
XM_017001877.1 | 2155 | Missense Mutation | CAA,GAA | Q,E 583 | XP_016857366.1 | |
XM_017001878.1 | 2155 | Missense Mutation | CAA,GAA | Q,E 573 | XP_016857367.1 | |
XM_017001879.1 | 2155 | Missense Mutation | CAA,GAA | Q,E 583 | XP_016857368.1 | |
XM_017001880.1 | 2155 | Missense Mutation | CAA,GAA | Q,E 544 | XP_016857369.1 | |
XM_017001881.1 | 2155 | Missense Mutation | CAA,GAA | Q,E 413 | XP_016857370.1 | |
XM_017001882.1 | 2155 | Missense Mutation | CAA,GAA | Q,E 308 | XP_016857371.1 |