Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATCGAGCTAGTGCCTTGAAGTCTC[C/T]GGAACAGCTGTGGGAGCCAAGAATC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603286 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GOLT1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GOLT1A - golgi transport 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198447.1 | 554 | Missense Mutation | CAG,CGG | Q,R 123 | NP_940849.1 | |
XM_017000314.1 | 554 | Intron | XP_016855803.1 |
KISS1 - KiSS-1 metastasis-suppressor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |