Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTAGGCCATGTAGAAGTCCATAGAG[A/G]TTGACCAATCGCTGTAACGCCTCCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 607560 MIM: 609043 | ||||||||||||||||||||
Literature Links: |
ARHGEF2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ARHGEF2 - Rho/Rac guanine nucleotide exchange factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001162383.1 | 2394 | Silent Mutation | AAC,AAT | N,N 744 | NP_001155855.1 | |
NM_001162384.1 | 2394 | Silent Mutation | AAC,AAT | N,N 743 | NP_001155856.1 | |
NM_004723.3 | 2394 | Silent Mutation | AAC,AAT | N,N 716 | NP_004714.2 | |
XM_005245587.2 | 2394 | Silent Mutation | AAC,AAT | N,N 744 | XP_005245644.1 | |
XM_005245588.2 | 2394 | Silent Mutation | AAC,AAT | N,N 743 | XP_005245645.1 | |
XM_005245589.4 | 2394 | Silent Mutation | AAC,AAT | N,N 736 | XP_005245646.1 | |
XM_005245590.3 | 2394 | Silent Mutation | AAC,AAT | N,N 729 | XP_005245647.1 | |
XM_005245591.3 | 2394 | Silent Mutation | AAC,AAT | N,N 729 | XP_005245648.1 | |
XM_005245592.2 | 2394 | Silent Mutation | AAC,AAT | N,N 717 | XP_005245649.1 | |
XM_005245593.4 | 2394 | Silent Mutation | AAC,AAT | N,N 717 | XP_005245650.1 | |
XM_005245594.2 | 2394 | Silent Mutation | AAC,AAT | N,N 717 | XP_005245651.1 | |
XM_006711622.3 | 2394 | Silent Mutation | AAC,AAT | N,N 727 | XP_006711685.1 | |
XM_006711623.3 | 2394 | Silent Mutation | AAC,AAT | N,N 668 | XP_006711686.1 | |
XM_006711624.3 | 2394 | Silent Mutation | AAC,AAT | N,N 668 | XP_006711687.1 | |
XM_006711625.3 | 2394 | Silent Mutation | AAC,AAT | N,N 668 | XP_006711688.1 | |
XM_006711626.2 | 2394 | Silent Mutation | AAC,AAT | N,N 668 | XP_006711689.1 | |
XM_011510136.2 | 2394 | Silent Mutation | AAC,AAT | N,N 886 | XP_011508438.2 | |
XM_011510137.2 | 2394 | Silent Mutation | AAC,AAT | N,N 887 | XP_011508439.2 | |
XM_011510139.2 | 2394 | Silent Mutation | AAC,AAT | N,N 887 | XP_011508441.1 | |
XM_011510141.2 | 2394 | Silent Mutation | AAC,AAT | N,N 740 | XP_011508443.1 | |
XM_011510143.2 | 2394 | Silent Mutation | AAC,AAT | N,N 565 | XP_011508445.1 | |
XM_017002794.1 | 2394 | Silent Mutation | AAC,AAT | N,N 886 | XP_016858283.1 | |
XM_017002795.1 | 2394 | Silent Mutation | AAC,AAT | N,N 735 | XP_016858284.1 | |
XM_017002796.1 | 2394 | Silent Mutation | AAC,AAT | N,N 726 | XP_016858285.1 | |
XM_017002797.1 | 2394 | Silent Mutation | AAC,AAT | N,N 716 | XP_016858286.1 | |
XM_017002798.1 | 2394 | Silent Mutation | AAC,AAT | N,N 668 | XP_016858287.1 | |
XM_017002799.1 | 2394 | Silent Mutation | AAC,AAT | N,N 667 | XP_016858288.1 |
MIR6738 - microRNA 6738 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RXFP4 - relaxin/insulin like family peptide receptor 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |