Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAAATAGGAAGAGAATAGTATTAC[C/T]CACCTCATAGGGTGGTTGTGAGGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602636 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP1R8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPP1R8 - protein phosphatase 1 regulatory subunit 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002713.3 | Intron | NP_002704.1 | ||||
NM_014110.4 | Intron | NP_054829.2 | ||||
NM_138558.2 | Intron | NP_612568.1 | ||||
XM_017001594.1 | Intron | XP_016857083.1 |
SCARNA1 - small Cajal body-specific RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |