Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGTGTGCAACAGTTGCGGCCTGGA[A/G]ATCGTGGACAAATACCTTCTCAAGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604425 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LHX8 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LHX8 - LIM homeobox 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001933.1 | 298 | Silent Mutation | GAA,GAG | E,E 81 | NP_001001933.1 | |
NM_001256114.1 | 298 | Silent Mutation | GAA,GAG | E,E 71 | NP_001243043.1 | |
XM_017001316.1 | 298 | Silent Mutation | GAA,GAG | E,E 71 | XP_016856805.1 | |
XM_017001317.1 | 298 | Silent Mutation | GAA,GAG | E,E 71 | XP_016856806.1 |