Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGACCTTGTCCGAGCGTCTATGC[A/G]CAGGAAGAAGAGCACCAGGGGTGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 604468 MIM: 608042 | ||||||||||||||||||||
Literature Links: |
MAP3K6 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAP3K6 - mitogen-activated protein kinase kinase kinase 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SYTL1 - synaptotagmin like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193308.1 | 517 | Missense Mutation | CAC,CGC | H,R 107 | NP_001180237.1 | |
NM_032872.2 | 517 | Missense Mutation | CAC,CGC | H,R 107 | NP_116261.1 | |
XM_005246022.3 | 517 | Missense Mutation | CAC,CGC | H,R 139 | XP_005246079.1 | |
XM_006710990.1 | 517 | Missense Mutation | CAC,CGC | H,R 139 | XP_006711053.1 | |
XM_006710991.1 | 517 | UTR 5 | XP_006711054.1 |