Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCCATCATTATGCTGAGCCCGGGC[A/G]TGGAGAGTCAGCTCTACAAGGTGAG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 614549 | ||||||||||||||||||||
Literature Links: |
SERINC2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SERINC2 - serine incorporator 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199037.1 | 1142 | Missense Mutation | ATG,GTG | M,V 65 | NP_001185966.1 | |
NM_001199038.1 | 1142 | Missense Mutation | ATG,GTG | M,V 70 | NP_001185967.1 | |
NM_001199039.1 | 1142 | Missense Mutation | ATG,GTG | M,V 6 | NP_001185968.1 | |
NM_018565.3 | 1142 | Missense Mutation | ATG,GTG | M,V 65 | NP_061035.2 | |
NM_178865.4 | 1142 | Missense Mutation | ATG,GTG | M,V 61 | NP_849196.2 |