Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCACCCAGCGTGGCTGCAGAAGCTC[A/T]CTAAGCACAAGACACCAGCACAGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 601231 | ||||||||||||||||||||
Literature Links: |
ANGPTL7 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ANGPTL7 - angiopoietin like 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021146.3 | 379 | Missense Mutation | ACT,TCT | T,S 30 | NP_066969.1 | |
XM_017000004.1 | 379 | Missense Mutation | ACT,TCT | T,S 30 | XP_016855493.1 |
MTOR - mechanistic target of rapamycin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004958.3 | 379 | Intron | NP_004949.1 | |||
XM_005263438.2 | 379 | Intron | XP_005263495.1 | |||
XM_011541166.2 | 379 | Intron | XP_011539468.1 | |||
XM_017000900.1 | 379 | Intron | XP_016856389.1 | |||
XM_017000901.1 | 379 | Intron | XP_016856390.1 | |||
XM_017000902.1 | 379 | Intron | XP_016856391.1 |