Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
GGGCTCCTGCTCCCACCCCTCCGGC[A/C]CCCCCACCGTCGCGCTCCTCCAGGC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 608863 | ||||||||||||||||||||
Literature Links: |
PDPN PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PDPN - podoplanin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001006624.1 | 163 | Intron | NP_001006625.1 | |||
NM_001006625.1 | 163 | Intron | NP_001006626.1 | |||
NM_006474.4 | 163 | Silent Mutation | GCA,GCC | A,A 38 | NP_006465.3 | |
NM_198389.2 | 163 | Silent Mutation | GCA,GCC | A,A 38 | NP_938203.2 | |
XM_006710295.1 | 163 | Intron | XP_006710358.1 |