Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGTGTCTCCTTGATGACCTGCGGC[A/G]ACGTCCAGCGCAGGATGTAGTGTGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 606811 | ||||||||||||||||||||
Literature Links: |
ALDH4A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ALDH4A1 - aldehyde dehydrogenase 4 family member A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001161504.1 | 1904 | Missense Mutation | TCG,TTG | S,L 483 | NP_001154976.1 | |
NM_001319218.1 | 1904 | Missense Mutation | TCG,TTG | S,L 492 | NP_001306147.1 | |
NM_003748.3 | 1904 | Missense Mutation | TCG,TTG | S,L 543 | NP_003739.2 | |
NM_170726.2 | 1904 | Missense Mutation | TCG,TTG | S,L 543 | NP_733844.1 |