Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAGAAGGAAAAGAAGCTTCCCCC[A/G]CCACCCCCTCAAGCCCCACCTGAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 609132 | ||||||||||||||||||||
Literature Links: |
KDM1A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
KDM1A - lysine demethylase 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009999.2 | 613 | Silent Mutation | CCA,CCG | P,P 153 | NP_001009999.1 | |
NM_015013.3 | 613 | Silent Mutation | CCA,CCG | P,P 153 | NP_055828.2 | |
XM_005245786.2 | 613 | Silent Mutation | CCA,CCG | P,P 153 | XP_005245843.1 | |
XM_006710472.3 | 613 | Silent Mutation | CCA,CCG | P,P 153 | XP_006710535.1 | |
XM_006710473.2 | 613 | Silent Mutation | CCA,CCG | P,P 153 | XP_006710536.1 | |
XM_006710474.3 | 613 | Silent Mutation | CCA,CCG | P,P 153 | XP_006710537.1 | |
XM_017000716.1 | 613 | Silent Mutation | CCA,CCG | P,P 153 | XP_016856205.1 | |
XM_017000717.1 | 613 | Silent Mutation | CCA,CCG | P,P 153 | XP_016856206.1 | |
XM_017000718.1 | 613 | UTR 5 | XP_016856207.1 | |||
XM_017000719.1 | 613 | UTR 5 | XP_016856208.1 | |||
XM_017000720.1 | 613 | UTR 5 | XP_016856209.1 |