Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGACCACTCGGTGAGCCATTTACAA[C/G]GCATATGTATCTTTTTTTTGTTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611609 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SIPA1L2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SIPA1L2 - signal induced proliferation associated 1 like 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020808.4 | 6323 | UTR 3 | NP_065859.3 | |||
XM_005273211.2 | 6323 | UTR 3 | XP_005273268.1 | |||
XM_005273212.4 | 6323 | UTR 3 | XP_005273269.1 | |||
XM_005273213.4 | 6323 | UTR 3 | XP_005273270.1 | |||
XM_011544243.2 | 6323 | UTR 3 | XP_011542545.1 | |||
XM_017001896.1 | 6323 | UTR 3 | XP_016857385.1 | |||
XM_017001897.1 | 6323 | UTR 3 | XP_016857386.1 | |||
XM_017001898.1 | 6323 | UTR 3 | XP_016857387.1 |