Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAGATCTTGCCCTTTTGAAGTTCT[C/G]AGGGGAGGTGTAGGGTGTCCACGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 139380 MIM: 611616 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GNB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GNB1 - G protein subunit beta 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282538.1 | 1434 | UTR 3 | NP_001269467.1 | |||
NM_001282539.1 | 1434 | UTR 3 | NP_001269468.1 | |||
NM_002074.4 | 1434 | UTR 3 | NP_002065.1 | |||
XM_017001059.1 | 1434 | UTR 3 | XP_016856548.1 | |||
XM_017001060.1 | 1434 | UTR 3 | XP_016856549.1 | |||
XM_017001061.1 | 1434 | UTR 3 | XP_016856550.1 |
NADK - NAD kinase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |