Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAGAACGAAGGAAAGATGAGGTG[A/G]GACAGCGCCGGGTATCCATGTAATC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 609738 MIM: 604634 | ||||||||||||||||||||
Literature Links: |
IGSF9 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IGSF9 - immunoglobulin superfamily member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135050.1 | 3083 | Missense Mutation | CCC,CTC | P,L 962 | NP_001128522.1 | |
NM_020789.3 | 3083 | Missense Mutation | CCC,CTC | P,L 946 | NP_065840.2 | |
XM_017001884.1 | 3083 | Intron | XP_016857373.1 |
TAGLN2 - transgelin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |