Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTGGAGTCTCTAAATTTGCGCCAG[C/T]TTGCCTGCAGAGCCACAGCAGTTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
GCSAML PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GCSAML - germinal center associated signaling and motility like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001281834.1 | 1310 | Intron | NP_001268763.1 | |||
NM_001281835.1 | 1310 | Intron | NP_001268764.1 | |||
NM_001281836.1 | 1310 | Intron | NP_001268765.1 | |||
NM_001281837.1 | 1310 | Intron | NP_001268766.1 | |||
NM_001281838.1 | 1310 | Intron | NP_001268767.1 | |||
NM_001281853.1 | 1310 | Intron | NP_001268782.1 | |||
NM_145278.4 | 1310 | Intron | NP_660321.1 | |||
XM_017000399.1 | 1310 | Intron | XP_016855888.1 |
GCSAML-AS1 - GCSAML antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
OR2C3 - olfactory receptor family 2 subfamily C member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198074.4 | 1310 | Silent Mutation | AAA,AAG | K,K 316 | NP_932340.3 |