Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACCATCATCTTTGTCCCCACCAG[A/C]CATAGGATCATGGTTGGGTGTGGTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
17 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615689 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AMIGO1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AMIGO1 - adhesion molecule with Ig like domain 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020703.2 | 1673 | Missense Mutation | GCT,TCT | A,S 429 | NP_065754.2 | |
XM_011541812.2 | 1673 | Missense Mutation | GCT,TCT | A,S 429 | XP_011540114.1 |
CYB561D1 - cytochrome b561 family member D1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |