Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCAGCTGCTTGCGCGGCGCCTTGCC[A/C]CCCGTTGACTTGCGCGCAGTCTGCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 602820 MIM: 606123 | ||||||||||||||||||||
Literature Links: |
HIST3H3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HIST3H3 - histone cluster 3, H3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003493.2 | 39 | Silent Mutation | GGG,GGT | G,G 13 | NP_003484.1 |
TRIM17 - tripartite motif containing 17 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |