Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGCCCACCAAGCTAATAATATCAC[A/G]GTAACAATGGCAGCTAATTTTACAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612682 MIM: 604094 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DRAXIN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DRAXIN - dorsal inhibitory axon guidance protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198545.3 | Intron | NP_940947.3 | ||||
XM_011541405.2 | Intron | XP_011539707.1 | ||||
XM_017001214.1 | Intron | XP_016856703.1 | ||||
XM_017001215.1 | Intron | XP_016856704.1 |
MAD2L2 - MAD2 mitotic arrest deficient-like 2 (yeast) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127325.1 | Intron | NP_001120797.1 | ||||
NM_006341.3 | Intron | NP_006332.3 | ||||
XM_011540507.1 | Intron | XP_011538809.1 |