Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTTCAGAAGAGAACGTACCTTAAA[C/T]CTTTCTCTAAATAGTTTTTCTTCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 600497 | ||||||||||||||||||||
Literature Links: |
C1orf168 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C1orf168 - chromosome 1 open reading frame 168 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001004303.4 | 2050 | Silent Mutation | AGA,AGG | R,R 656 | NP_001004303.3 | |
XM_005270584.3 | 2050 | Silent Mutation | AGA,AGG | R,R 548 | XP_005270641.1 | |
XM_011540898.1 | 2050 | Silent Mutation | AGA,AGG | R,R 682 | XP_011539200.1 | |
XM_011540899.1 | 2050 | Silent Mutation | AGA,AGG | R,R 652 | XP_011539201.1 | |
XM_011540900.2 | 2050 | Intron | XP_011539202.1 | |||
XM_011540903.2 | 2050 | Intron | XP_011539205.1 | |||
XM_011540904.2 | 2050 | Silent Mutation | AGA,AGG | R,R 374 | XP_011539206.1 | |
XM_011540905.2 | 2050 | Intron | XP_011539207.1 | |||
XM_011540906.2 | 2050 | Silent Mutation | AGA,AGG | R,R 312 | XP_011539208.1 | |
XM_017000544.1 | 2050 | Silent Mutation | AGA,AGG | R,R 526 | XP_016856033.1 | |
XM_017000545.1 | 2050 | Silent Mutation | AGA,AGG | R,R 526 | XP_016856034.1 | |
XM_017000546.1 | 2050 | Intron | XP_016856035.1 | |||
XM_017000547.1 | 2050 | Intron | XP_016856036.1 | |||
XM_017000548.1 | 2050 | Intron | XP_016856037.1 |
PRKAA2 - protein kinase AMP-activated catalytic subunit alpha 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |