Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATGCCTGTGGCCAAGGGTGCTCTC[C/T]GGATCCCGCCTGAGTGTGGATCCGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608205 MIM: 602454 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MECR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MECR - mitochondrial trans-2-enoyl-CoA reductase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PTPRU - protein tyrosine phosphatase, receptor type U | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195001.1 | Intron | NP_001181930.1 | ||||
NM_005704.4 | Intron | NP_005695.3 | ||||
NM_133177.3 | Intron | NP_573438.3 | ||||
NM_133178.3 | Intron | NP_573439.2 | ||||
XM_006710269.2 | Intron | XP_006710332.1 | ||||
XM_016999992.1 | Intron | XP_016855481.1 | ||||
XM_016999993.1 | Intron | XP_016855482.1 | ||||
XM_016999994.1 | Intron | XP_016855483.1 | ||||
XM_016999995.1 | Intron | XP_016855484.1 |