Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGATCACCTTTGTGGATTTCCTT[A/G]CCTATGATGTCCTTGACATGAAGCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 138385 | ||||||||||||||||||||
Literature Links: |
GSTM5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GSTM5 - glutathione S-transferase mu 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000851.3 | 575 | Missense Mutation | ACC,GCC | T,A 160 | NP_000842.2 | |
XM_005270784.4 | 575 | Missense Mutation | ACC,GCC | T,A 160 | XP_005270841.1 | |
XM_005270785.4 | 575 | Missense Mutation | ACC,GCC | T,A 56 | XP_005270842.1 |