Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTTAACAACACTGGTATTATTAG[A/T]GAGCTGATAATGAAACTAAGAATGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604388 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GNG4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GNG4 - G protein subunit gamma 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098721.1 | 2695 | UTR 3 | NP_001092191.1 | |||
NM_001098722.1 | 2695 | UTR 3 | NP_001092192.1 | |||
NM_004485.3 | 2695 | UTR 3 | NP_004476.1 | |||
XM_006711761.2 | 2695 | UTR 3 | XP_006711824.1 | |||
XM_011544167.2 | 2695 | UTR 3 | XP_011542469.1 |