Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCAGCATCTTGATGAACCACTCGCC[A/G]TCGCGTGGGCAGGGGGATGCGCGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611413 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DLGAP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DLGAP3 - DLG associated protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080418.2 | 2360 | Silent Mutation | GAC,GAT | D,D 794 | NP_001073887.1 | |
XM_011541879.2 | 2360 | Silent Mutation | GAC,GAT | D,D 794 | XP_011540181.1 | |
XM_011541880.2 | 2360 | Silent Mutation | GAC,GAT | D,D 297 | XP_011540182.1 |
SMIM12 - small integral membrane protein 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |