Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATTCCATCATAATAATCCCGTTG[A/C]AAGCCATAGTCCAAGTCAAAAGAGG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
6 submissions
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
HNRNPCL1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
HNRNPCL1 - heterogeneous nuclear ribonucleoprotein C-like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013631.2 | Intron | NP_001013653.1 |
PRAMEF2 - PRAME family member 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |