Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCAGTAAAACATCACTGTATTACT[A/G]TTTTACTTGGGAAGAAAAACATAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 147880 MIM: 610482 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL6R PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
IL6R - interleukin 6 receptor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SHE - Src homology 2 domain containing E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010846.2 | Intron | NP_001010846.1 | ||||
XM_005244891.4 | Intron | XP_005244948.1 | ||||
XM_011509163.2 | Intron | XP_011507465.1 | ||||
XM_017000256.1 | Intron | XP_016855745.1 |