Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TATTCAGTTGACAGCGCCAGGTGCT[A/G]GTGACTATAAGGTGATGGGTTCAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
14 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
PAPPA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PAPPA2 - pappalysin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_020318.2 | Intron | NP_064714.2 | ||||
NM_021936.2 | Intron | NP_068755.2 | ||||
XM_005245422.3 | Intron | XP_005245479.1 | ||||
XM_011509857.1 | Intron | XP_011508159.1 | ||||
XM_011509858.2 | Intron | XP_011508160.1 | ||||
XM_017002023.1 | Intron | XP_016857512.1 | ||||
XM_017002024.1 | Intron | XP_016857513.1 | ||||
XM_017002025.1 | Intron | XP_016857514.1 |