Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGGGGCAGTGAAGCAGGGCGGTG[A/G]GCATGGCCAGTGGTGGGCTTCGGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
15 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 180231 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CRABP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CRABP2 - cellular retinoic acid binding protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199723.1 | 684 | UTR 3 | NP_001186652.1 | |||
NM_001878.3 | 684 | UTR 3 | NP_001869.1 | |||
XM_017000343.1 | 684 | UTR 3 | XP_016855832.1 |