Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTATATTCTTTGATGATTTCTTCT[A/C]TTTTCCTGCAGTCTCTTTTTAAGCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610860 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AGL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AGL - amylo-alpha-1, 6-glucosidase, 4-alpha-glucanotransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000028.2 | Intron | NP_000019.2 | ||||
NM_000642.2 | Intron | NP_000633.2 | ||||
NM_000643.2 | Intron | NP_000634.2 | ||||
NM_000644.2 | Intron | NP_000635.2 | ||||
NM_000645.2 | Intron | NP_000636.2 | ||||
NM_000646.2 | Intron | NP_000637.2 | ||||
XM_005270557.2 | Intron | XP_005270614.1 | ||||
XM_017000501.1 | Intron | XP_016855990.1 |