Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGTAAAGAAGGCTAGGAAAACTTA[A/C]GAAGCATGGGGCATTTGAGTTGGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
17 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610617 MIM: 611350 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DTL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DTL - denticleless E3 ubiquitin protein ligase homolog | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286229.1 | Intron | NP_001273158.1 | ||||
NM_001286230.1 | Intron | NP_001273159.1 | ||||
NM_016448.3 | Intron | NP_057532.3 | ||||
XM_011509614.1 | Intron | XP_011507916.1 |
INTS7 - integrator complex subunit 7 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPL21P28 - ribosomal protein L21 pseudogene 28 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |