Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTACTTTGTCCTCGGGGTTCCAA[C/T]GGAGTTTCTGTTCTAAATCAGTCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
7 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613360 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DRAM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DRAM2 - DNA damage regulated autophagy modulator 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_178454.4 | 1019 | Missense Mutation | CAT,CGT | H,R 194 | NP_848549.3 | |
XM_005270469.2 | 1019 | Missense Mutation | CAT,CGT | H,R 194 | XP_005270526.1 | |
XM_005270470.1 | 1019 | Missense Mutation | CAT,CGT | H,R 194 | XP_005270527.1 | |
XM_006710361.1 | 1019 | Missense Mutation | CAT,CGT | H,R 104 | XP_006710424.1 | |
XM_006710362.1 | 1019 | Missense Mutation | CAT,CGT | H,R 104 | XP_006710425.1 | |
XM_011540707.1 | 1019 | Missense Mutation | CAT,CGT | H,R 194 | XP_011539009.1 | |
XM_011540708.1 | 1019 | Missense Mutation | CAT,CGT | H,R 194 | XP_011539010.1 | |
XM_017000320.1 | 1019 | Missense Mutation | CAT,CGT | H,R 104 | XP_016855809.1 | |
XM_017000321.1 | 1019 | Missense Mutation | CAT,CGT | H,R 104 | XP_016855810.1 |