Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTTTGTGTGACAGCAGGACATCCG[A/G]GTTTGGAGCATCCTCCACCACCCAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM129A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FAM129A - family with sequence similarity 129 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_052966.3 | 3695 | Missense Mutation | CCG,TCG | P,S 901 | NP_443198.1 | |
XM_011509140.2 | 3695 | Missense Mutation | CCG,TCG | P,S 960 | XP_011507442.1 | |
XM_011509141.2 | 3695 | Missense Mutation | CCG,TCG | P,S 923 | XP_011507443.1 |