Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTTATTTTCTTCTTCAGTGGGCCA[C/T]TGTCCTGATCCTGTGCTGGTGAATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 120831 | ||||||||||||||||||||
Literature Links: |
C4BPB PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C4BPB - complement component 4 binding protein beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000716.3 | 530 | Silent Mutation | CAC,CAT | H,H 80 | NP_000707.1 | |
NM_001017364.1 | 530 | Silent Mutation | CAC,CAT | H,H 79 | NP_001017364.1 | |
NM_001017365.2 | 530 | Silent Mutation | CAC,CAT | H,H 80 | NP_001017365.1 | |
NM_001017366.2 | 530 | Silent Mutation | CAC,CAT | H,H 79 | NP_001017366.1 | |
NM_001017367.1 | 530 | Silent Mutation | CAC,CAT | H,H 80 | NP_001017367.1 | |
XM_005273254.4 | 530 | Silent Mutation | CAC,CAT | H,H 80 | XP_005273311.1 | |
XM_005273255.2 | 530 | Silent Mutation | CAC,CAT | H,H 79 | XP_005273312.1 |