Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGCTTAGGAAGTTTAAGTAACTT[C/T]CACAAGGTCACATAGTTAGTCAATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
11 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606080 MIM: 614234 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHIA PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHIA - chitinase, acidic | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PIFO - primary cilia formation | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001300831.1 | Intron | NP_001287760.1 | ||||
NM_181643.5 | Intron | NP_857594.2 | ||||
XM_005270472.1 | Intron | XP_005270529.1 | ||||
XM_017000322.1 | Intron | XP_016855811.1 |