Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCGCAGGTTTCTGCCACCTCTTTCT[A/G]CCAGATGAACAGTTTCCTGAAGGGT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 182310 | ||||||||||||||||||||
Literature Links: |
ATP1A1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATP1A1 - ATPase Na+/K+ transporting subunit alpha 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000701.7 | 2043 | Silent Mutation | CTA,CTG | L,L 560 | NP_000692.2 | |
NM_001160233.1 | 2043 | Silent Mutation | CTA,CTG | L,L 560 | NP_001153705.1 | |
NM_001160234.1 | 2043 | Silent Mutation | CTA,CTG | L,L 529 | NP_001153706.1 | |
XM_017001360.1 | 2043 | Silent Mutation | CTA,CTG | L,L 529 | XP_016856849.1 | |
XM_017001361.1 | 2043 | Silent Mutation | CTA,CTG | L,L 529 | XP_016856850.1 |
ATP1A1-AS1 - ATP1A1 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |