Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCCACTTGCCATCAGTGCTCCCAC[A/G]ATGACCGAGTAAGCAATTACACTAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TTC13 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TTC13 - tetratricopeptide repeat domain 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001122835.2 | 3380 | Silent Mutation | ATC,ATT | I,I 723 | NP_001116307.2 | |
NM_024525.4 | 3380 | Silent Mutation | ATC,ATT | I,I 777 | NP_078801.3 | |
XM_005273260.1 | 3380 | Silent Mutation | ATC,ATT | I,I 776 | XP_005273317.1 | |
XM_005273261.2 | 3380 | Silent Mutation | ATC,ATT | I,I 746 | XP_005273318.1 | |
XM_005273262.1 | 3380 | Silent Mutation | ATC,ATT | I,I 724 | XP_005273319.1 | |
XM_005273264.2 | 3380 | Silent Mutation | ATC,ATT | I,I 701 | XP_005273321.1 | |
XM_006711814.2 | 3380 | Silent Mutation | ATC,ATT | I,I 648 | XP_006711877.1 | |
XM_006711815.2 | 3380 | Silent Mutation | ATC,ATT | I,I 419 | XP_006711878.1 | |
XM_011544275.2 | 3380 | Silent Mutation | ATC,ATT | I,I 670 | XP_011542577.1 | |
XM_011544276.2 | 3380 | Silent Mutation | ATC,ATT | I,I 480 | XP_011542578.1 | |
XM_017002321.1 | 3380 | Silent Mutation | ATC,ATT | I,I 745 | XP_016857810.1 | |
XM_017002322.1 | 3380 | Silent Mutation | ATC,ATT | I,I 700 | XP_016857811.1 | |
XM_017002323.1 | 3380 | Silent Mutation | ATC,ATT | I,I 648 | XP_016857812.1 | |
XM_017002324.1 | 3380 | Silent Mutation | ATC,ATT | I,I 647 | XP_016857813.1 | |
XM_017002325.1 | 3380 | Silent Mutation | ATC,ATT | I,I 418 | XP_016857814.1 | |
XM_017002326.1 | 3380 | Silent Mutation | ATC,ATT | I,I 418 | XP_016857815.1 |