Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTCATTGCCTACTTCCTCATCTGG[C/T]TCGTGCCCGACTTCCCACACGGCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 614397 | ||||||||||||||||||||
Literature Links: |
MFSD2A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MFSD2A - major facilitator superfamily domain containing 2A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136493.2 | 473 | Missense Mutation | CTC,TTC | L,F 148 | NP_001129965.1 | |
NM_001287808.1 | 473 | UTR 5 | NP_001274737.1 | |||
NM_001287809.1 | 473 | Missense Mutation | CTC,TTC | L,F 98 | NP_001274738.1 | |
NM_032793.4 | 473 | Missense Mutation | CTC,TTC | L,F 135 | NP_116182.2 | |
XM_005271285.1 | 473 | Missense Mutation | CTC,TTC | L,F 133 | XP_005271342.1 | |
XM_011542312.2 | 473 | Missense Mutation | CTC,TTC | L,F 135 | XP_011540614.1 |