Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCAGCAGTTCCCGCAGTTCCACGT[C/G]AAGTCCGGCCTGCAGATCAAGAAGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
2 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 602006 | ||||||||||||||||||||
Literature Links: |
MAPKAPK2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
MAPKAPK2 - mitogen-activated protein kinase-activated protein kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004759.4 | 354 | Silent Mutation | GTC,GTG | V,V 48 | NP_004750.1 | |
NM_032960.3 | 354 | Silent Mutation | GTC,GTG | V,V 48 | NP_116584.2 | |
XM_005273353.3 | 354 | Silent Mutation | GTC,GTG | V,V 48 | XP_005273410.1 | |
XM_017002810.1 | 354 | Intron | XP_016858299.1 |