Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTAATAAAATCCCTCAAACAGCA[C/G]TTTTCTACTTGTTTCGGAGTACACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603941 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC181 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC181 - coiled-coil domain containing 181 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC19A2 - solute carrier family 19 member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319667.1 | 2709 | UTR 3 | NP_001306596.1 | |||
NM_006996.2 | 2709 | UTR 3 | NP_008927.1 |