Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTGGCGGCCCCCACTCTAGTGCT[G/A]TGGTAGGACAATGAGCTCACAGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
9 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602250 MIM: 604097 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TNFRSF9 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TNFRSF9 - TNF receptor superfamily member 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001561.5 | 2071 | UTR 3 | NP_001552.2 | |||
XM_006710618.3 | 2071 | Intron | XP_006710681.1 | |||
XM_011541386.2 | 2071 | UTR 3 | XP_011539688.1 |
UTS2 - urotensin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |