Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGGACAGTAGGATCTTTCGAAGT[A/G]AGGTATTAATGCAGTCAAACCTTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
3 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 184755 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ECHDC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ECHDC2 - enoyl-CoA hydratase domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR1273F - microRNA 1273f | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR5095 - microRNA 5095 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCP2 - sterol carrier protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007098.2 | Intron | NP_001007099.1 | ||||
NM_001007099.2 | Intron | NP_001007100.1 | ||||
NM_001007100.2 | Intron | NP_001007101.1 | ||||
NM_001007250.2 | Intron | NP_001007251.1 | ||||
NM_001193599.1 | Intron | NP_001180528.1 | ||||
NM_001193600.1 | Intron | NP_001180529.1 | ||||
NM_001193617.1 | Intron | NP_001180546.1 | ||||
NM_002979.4 | Intron | NP_002970.2 | ||||
XM_005271103.4 | Intron | XP_005271160.1 | ||||
XM_011541935.2 | Intron | XP_011540237.1 | ||||
XM_017002045.1 | Intron | XP_016857534.1 | ||||
XM_017002046.1 | Intron | XP_016857535.1 |