Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGCTGCCGGGTTGCGTTCTGGGG[A/G]TCGCCCTGCTCCCCCTCCTCCTCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
8 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611321 MIM: 602839 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLSTN1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLSTN1 - calsyntenin 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009566.2 | 3679 | Silent Mutation | NP_001009566.1 | |||
NM_001302883.1 | 3679 | Silent Mutation | NP_001289812.1 | |||
NM_014944.4 | 3679 | Silent Mutation | NP_055759.3 |
PIK3CD - phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit delta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |